missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)propionic acid, 96%

Product Code. 15436517
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436517 50 g 50g
15426517 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436517 Supplier Thermo Scientific Alfa Aesar Supplier No. H33663.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16642-79-8 | C9H9NO4 | 195.17 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16642-79-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00126834
InChI Key VZOPVJNBOQOLPN-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
PubChem CID 85526
IUPAC Name 3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 164°C to 168°C
Quantity 50 g
Solubility Information Slightly soluble in water.
Formula Weight 195.17
Percent Purity 96%
Chemical Name or Material 3-(4-Nitrophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.