missing translation for 'onlineSavingsMsg'
Learn More

3-(4-n-Propoxyphenyl)propionic acid, 96%

Product Code. 15486487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486487 1 g 1g
15496487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486487 Supplier Thermo Scientific Alfa Aesar Supplier No. H33646.03
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3243-40-1 | C12H16O3 | 208.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3243-40-1
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00484121
InChI Key SOFMLHAGBATXNZ-UHFFFAOYSA-N
Synonym 3-4-propoxyphenyl propanoic acid, 3-4-propoxy-phenyl-propionic acid, benzenepropanoic acid,4-propoxy, 3-4-n-propoxyphenyl propionic acid, cbmicro_011397, cambridge id 5152838, 4-propoxy-benzenepropanoic acid, 3-4-n-propoxyphenyl propanoic acid
PubChem CID 2255935
IUPAC Name 3-(4-propoxyphenyl)propanoic acid
SMILES CCCOC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 94°C to 99°C
Quantity 1 g
Formula Weight 208.26
Percent Purity 96%
Chemical Name or Material 3-(4-n-Propoxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.