missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propyl isothiocyanate, 96%, Thermo Scientific Chemicals

Catalog Number p-7046017
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
32813-50-6
C8H14N2OS
186.27
MFCD00041222
BCEFDMYFAAAFPE-UHFFFAOYSA-N
3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
141745
4-(3-isothiocyanatopropyl)morpholine
S=C=NCCCN1CCOCC1
This item is not returnable. View return policy
32813-50-6
C8H14N2OS
186.27
MFCD00041222
BCEFDMYFAAAFPE-UHFFFAOYSA-N
3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
141745
4-(3-isothiocyanatopropyl)morpholine
S=C=NCCCN1CCOCC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 182°C to 183°C (13 mmHg)
Refractive Index 1.5305
Quantity 1 g
UN Number UN2922
Beilstein 879217
Sensitivity Moisture sensitive
Formula Weight 186.28
Percent Purity 96%
Chemical Name or Material 3-(4-Morpholinyl)propyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'