missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)-1-propanol, 95%

Product Code. 15446838
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446838 5 g 5g
15436838 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446838 Supplier Thermo Scientific Alfa Aesar Supplier No. H60376.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4441-30-9 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4441-30-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD01713533
InChI Key VZKSLWJLGAGPIU-UHFFFAOYSA-N
Synonym 4-3-hydroxypropyl morpholine, 3-morpholinopropanol, 4-morpholinepropanol, 3-morpholin-4-yl propan-1-ol, 3-morpholinopropan-1-ol, 3-4-morpholinyl-1-propanol, 3-morpholin-4-yl-propan-1-ol, 3-morpholino-1-propanol, gamma-morpholinopropanol german, n-hydroxypropanylmorpholine
PubChem CID 199576
IUPAC Name 3-morpholin-4-ylpropan-1-ol
SMILES C1COCCN1CCCO

Specifications

Density 1.049
Boiling Point 240°C to 241°C
Flash Point 85°C (185°F)
Refractive Index 1.477
Quantity 5 g
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 3-(4-Morpholinyl)-1-propanol
compliance-icons
Product Identifier
  • 3-(4-Morpholinyl)-1-propanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Morpholinopropanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.