missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylpiperazin-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10341053
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341053 1 g Amber Glass Bottle 1g
10750744 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341053 Supplier Thermo Scientific Maybridge Supplier No. CC35901DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 215309-01-6
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD06742238
InChI Key HALATUFUWLWCQV-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
PubChem CID 4741681
IUPAC Name 3-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 3-(4-Methylpiperazin-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.