Learn More
3-(4-Methylpiperazin-1-yl)aniline, 97%, Maybridge™
Brand: Maybridge CC35914CB
Code : 31
Additional Details : CAS Number : 148546-99-0 Weight : 0.05025kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
148546-99-0 | |
191.278 | |
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
3-(4-methylpiperazin-1-yl)aniline |
C11H17N3 | |
RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
11564613 | |
CN1CCN(CC1)C2=CC(=CC=C2)N |
Specifications
3-(4-Methylpiperazin-1-yl)aniline | |
250mg | |
97% | |
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
CN1CCN(CC1)C2=CC(=CC=C2)N | |
191.278 | |
191.28 |
Amber Glass Bottle | |
148546-99-0 | |
C11H17N3 | |
RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
3-(4-methylpiperazin-1-yl)aniline | |
11564613 | |
97% |