missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylphenyl)propionic acid, 98+%, Thermo Scientific™

Product Code. 11491858
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491858 5 g 5g
11389549 10 g 10g
11401868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491858 Supplier Thermo Scientific Alfa Aesar Supplier No. A14513.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(p-Tolyl)propionic acid was used during the coupling of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide with 4,5-diphenyl-2-(2-p-tolyl-ethyl)-4,5-dihydro-1H-imidazole resin.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1505-50-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00016561
InChI Key LDYGRLNSOKABMM-UHFFFAOYSA-N
Synonym 3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
PubChem CID 73927
IUPAC Name 3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 115°C to 117°C
Quantity 5 g
Beilstein 2046164
Solubility Information Soluble in methanol.
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 3-(4-Methylphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.