missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]benzylamine, 95%, Thermo Scientific™

Product Code. 11376795
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11376795 250 mg Amber Glass Bottle 250mg
10472674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376795 Supplier Thermo Scientific Maybridge Supplier No. CC55913CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-48-1
Molecular Formula C14H23N3
Molecular Weight (g/mol) 233.36
MDL Number MFCD09787493
InChI Key MSLZRDIUXASDJH-UHFFFAOYSA-N
Synonym 3-4-methylperhydro-1,4-diazepin-1-yl methyl benzylamine, 3-4-methylhomopiperazin-1-yl methyl benzylamine, 3-4-methyl-1,4-diazepan-1-yl methyl phenyl methanamine, 1-3-4-methyl-1,4-diazepan-1-yl methyl phenyl methanamine, 3-4-methyl-1,4-diazaperhydroepinyl methyl phenyl methylamine, benzenemethanamine,3-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl, 3-4-methylhomopiperazin-1-yl methyl benzylamine, 3-4-methyl-1,4-diazepan-1-yl methyl phenyl methylamine
PubChem CID 24229554
IUPAC Name [3-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl]methanamine
SMILES CN1CCCN(CC2=CC(CN)=CC=C2)CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 233.36
Percent Purity 95%
Chemical Name or Material 3-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.