missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylmagnesium bromide, 0.8M solution in toluene/THF (50:50), AcroSeal™

Product Code. 11975991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11975991 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11975991 Supplier Thermo Scientific Acros Supplier No. 438761000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17680-04-5 | C7H5BrMgO2 | 225.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17680-04-5, 109-99-9, 108-88-3
CAS Min % 15.17, 80.5
CAS Max % 19.5, 84.83
Color Colorless to Yellow
Density 1.0400g/mL
Flash Point −10°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C7H5BrMgO2
MDL Number MFCD01863705
Quantity 100 mL
Specific Gravity 1.04
Synonym 3,4-methylenedioxy phenylmagnesium bromide, jfuhzuguhblmlb-uhfffaoysa-m, 1,3-benzodioxol-5-yl magnesium bromide, 3,4-methylenedioxyphenyl magnesium bromide, 3.4-methylene dioxy-phenyl-magnesium bromide, 5-bromomagnesio-2h-1,3-benzodioxole, 3,4-methylenedioxyphenylmagnesium bromide, 3,4-methylenedioxy phenylmagnesium bromide, 0.50 m in 2-methf, 3,4-methylenedioxy phenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-methylenedioxy phenylmagnesium bromide solution, 1.0 m in thf: toluene 1:1, grignard reagent
InChI Key JFUHZUGUHBLMLB-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=C2OCOC2=C1
IUPAC Name magnesium;5H-1,3-benzodioxol-5-ide;bromide
Molecular Weight (g/mol) 225.32
PubChem CID 4429644
Formula Weight 225.32
Physical Form Liquid
Chemical Name or Material 3, 4-(Methylenedioxy)phenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,4-(Methylenedioxy)phenylmagnesium bromide, 1M solution in toluene/THF(50:50)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.