missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylmagnesium bromide, 0.50 M in 2-MeTHF

Product Code. 15401426
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15401426

Brand: Thermo Scientific Alfa Aesar H54632.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17680-04-5 | C7H5BrMgO2 | 225.32 g/mol

3,4-(Methylenedioxy)phenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agen
TRUSTED_SUSTAINABILITY

Specifications

CAS 17680-04-5
Molecular Formula C7H5BrMgO2
MDL Number MFCD01863705
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 3,4-methylenedioxy phenylmagnesium bromide, jfuhzuguhblmlb-uhfffaoysa-m, 1,3-benzodioxol-5-yl magnesium bromide, 3,4-methylenedioxyphenyl magnesium bromide, 3.4-methylene dioxy-phenyl-magnesium bromide, 5-bromomagnesio-2h-1,3-benzodioxole, 3,4-methylenedioxyphenylmagnesium bromide, 3,4-methylenedioxy phenylmagnesium bromide, 0.50 m in 2-methf, 3,4-methylenedioxy phenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-methylenedioxy phenylmagnesium bromide solution, 1.0 m in thf: toluene 1:1, grignard reagent
Solubility Information It reacts with water.
InChI Key JFUHZUGUHBLMLB-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=C2OCOC2=C1
IUPAC Name magnesium;5H-1,3-benzodioxol-5-ide;bromide
Molecular Weight (g/mol) 225.32
PubChem CID 4429644
Formula Weight 225.32
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3,4-(Methylenedioxy)phenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.