missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylacetonitrile, 98%, ACROS Organics™

Product Code. 10501462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10501462 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10501462 Supplier Thermo Scientific Acros Supplier No. 163350100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4439-02-5
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H7NO2
MDL Number MFCD00005835
Quantity 10 g
Synonym 3,4-methylenedioxy phenylacetonitrile, 1,3-benzodioxole-5-acetonitrile, 3,4-methylenedioxyphenylacetonitrile, 2-benzo d 1,3 dioxol-5-yl acetonitrile, 3,4-methylenedioxybenzyl cyanide, 2-2h-1,3-benzodioxol-5-yl acetonitrile, 1,3-benzodioxol-5-ylacetonitrile, 2-1,3-benzodioxol-5-yl acetonitrile, 3,4-methylenedioxybenzylcyanide, homopiperonylonitrile
InChI Key ZQPBOYASBNAXOZ-UHFFFAOYSA-N
SMILES C1OC2=C(O1)C=C(C=C2)CC#N
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetonitrile
Molecular Weight (g/mol) 161.16
PubChem CID 78178
Formula Weight 161.16
Percent Purity ≥97.5%
Physical Form Low Melting Solid
Chemical Name or Material 3, 4-(Methylenedioxy)phenylacetonitrile, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.