missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylacetic acid, 98.5+%

Product Code. 10137513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass bottle
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137513 25 g Glass bottle 25g
10512471 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10137513 Supplier Thermo Scientific Acros Supplier No. 302620250
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2861-28-1 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2861-28-1
MDL Number MFCD00014576
InChI Key ODVLMCWNGKLROU-UHFFFAOYSA-N
Synonym 1,3-benzodioxole-5-acetic acid, 3,4-methylenedioxy phenylacetic acid, 2-benzo d 1,3 dioxol-5-yl acetic acid, 3,4-methylenedioxyphenylacetic acid, homopiperonylic acid, benzo-1,3-dioxole-5-acetic acid, 1,3-benzodioxol-5-yl acetic acid, 2-2h-1,3-benzodioxol-5-yl acetic acid, 2-1,3-benzodioxol-5-yl acetic acid, 1,3-benzodioxol-5-ylacetic acid
PubChem CID 76115
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetic acid
SMILES C1OC2=C(O1)C=C(C=C2)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 132.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5+%
Packaging Glass bottle
Molecular Formula C9H8O4
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl alcohol
Molecular Weight (g/mol) 180.18
Formula Weight 180.18
Percent Purity 98.5+%
Physical Form Micro-crystalline Powder
Chemical Name or Material 3, 4-(Methylenedioxy)phenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.