missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzyl isothiocyanate, 94%, Thermo Scientific™

Product Code. 11388465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388465 1 g 1g
11398465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11388465 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L10501.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4430-47-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041217
InChI Key PUJWRDBPAFJUJW-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
PubChem CID 2795371
IUPAC Name 5-(isothiocyanatomethyl)-1,3-benzodioxole
SMILES S=C=NCC1=CC=C2OCOC2=C1

Specifikationer

Density 1.32
Boiling Point 139°C to 140°C (1 mmHg)
Odor Pungent
Refractive Index 1.615
Quantity 1 g
UN Number UN2922
Beilstein 1428941
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 193.23
Percent Purity 94%
Chemical Name or Material 3,4-(Methylenedioxy)benzyl isothiocyanate
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.