missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)aniline, 98+%

Product Code. 11459843
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11459843

Brand: Thermo Scientific Alfa Aesar A12975.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14268-66-7 | C7H7NO2 | 137.138 g/mol

3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong acids, acid chlorides, acid anhydrides. Air & Light Sensitive. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14268-66-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00005832
InChI Key XGNXYCFREOZBOL-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
PubChem CID 84310
IUPAC Name 1,3-benzodioxol-5-amine
SMILES C1OC2=C(O1)C=C(C=C2)N

Specifications

Melting Point 40°C to 45°C
Boiling Point 144°C (16 mmHg)
Flash Point 142°C (287°F)
Quantity 50 g
UN Number UN2811
Beilstein 4919
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.14
Percent Purity ≥98%
Chemical Name or Material 3,4-(Methylenedioxy)aniline
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.