missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole, Alfa Aesar™

Product Code. 11470170
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470170 250 mg 250mg
11480170 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470170 Supplier Thermo Scientific Alfa Aesar Supplier No. H50649.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 338393-13-8
Molecular Formula C10H11N3O2S
Molecular Weight (g/mol) 237.277
MDL Number MFCD01568307
InChI Key YYQTWXHTIYSIRV-UHFFFAOYSA-N
Synonym 3-4-methylbenzyl sulfonyl-1h-1,2,4-triazole, 4-methylbenzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methanesulfonyl-1h-1,2,4-triazole, acmc-20anb6, methylbenzyltriazolylsulfone, 3-p-tolylmethylsulfonyl-1h-1,2,4-triazole, 3-p-tolylmethanesulfonyl-1h-1,2,4 triazole, 4-methyl-benzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methyl sulfonyl-1,2,4-triazole
PubChem CID 2063879
IUPAC Name 5-[(4-methylphenyl)methylsulfonyl]-1H-1,2,4-triazole
SMILES CC1=CC=C(C=C1)CS(=O)(=O)C2=NC=NN2

Specifications

Quantity 250 mg
Formula Weight 237.28
Chemical Name or Material 3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.