missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline, TRC

Product Code. 30134147
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134147 250 mg 250mg
30009440 1 g 1g
30006222 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134147 Supplier Toronto Research Chemicals Supplier No. TRCM313065250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641571-11-1
Molecular Formula C11 H10 F3 N3
Molecular Weight (g/mol) 241.21
Synonym Benzenamine, 3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)-, 3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzenamine, 1-(3-Amino-5-trifluoromethylphenyl)-4-methylimidazole, 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline, 3-(4-Methylimidazol-1-yl)-5-trifluoromethylphenylamine, 5-(4-Methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)benzenamine, 5-(4-Methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)benzeneamine, Nilotinib Impurity
IUPAC Name 3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)aniline
SMILES Cc1cn(cn1)c2cc(N)cc(c2)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H10F3N3/c1-7-5-17(6-16-7)10-3-8(11(12,13)14)2-9(15)4-10/h2-6H,15H2,1H3
Formula Weight 241.0827
Chemical Name or Material 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.