missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10306424
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306424 5 g Amber Glass Bottle 5g
10732493 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306424 Supplier Thermo Scientific Maybridge Supplier No. MO00392DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 93041-45-3
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.22
MDL Number MFCD03407357
InChI Key IKDPECCMAKOJTK-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-5-methylisoxazole-4-carboxylic acid, 3-4-methoxyphenyl-5-methyl-4-isoxazolecarboxylic acid, 3-4-methoxyphenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylicacid, 3-4-methoxyphenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-methoxyphenyl-5-methyl, 3-4-methoxyphenyl-5-methyl-4-isoxazole carboxylic acid, 3-4-methoxyphenyl-5-methyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-4-methoxyphenyl-5-methyl
PubChem CID 2779764
IUPAC Name 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES COC1=CC=C(C=C1)C1=NOC(C)=C1C(O)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 233.22
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.