missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10496672
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10496672 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10496672 Supplier Thermo Scientific Maybridge Supplier No. SEW05757DA
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 199682-73-0
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C11H10N2O2
MDL Number MFCD01133696
Quantity 1 g
Synonym 3-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde, 5-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde, 3-4-methoxyphenyl-1h-pyrazole-4-carboxaldehyde, 3-4-methoxyphenyl pyrazole-4-carbaldehyde, 3-4-methoxyphenyl-2h-pyrazole-4-carbaldehyde, cbmicro_017678, cambridge id 6132430, 3-4-methoxyphenyl pyrazole-4-carboxaldehyde, 3-4-methoxy-phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carbaldehyde, 3-4-methoxyphenyl
InChI Key QSGGFCPKXTULQQ-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C2=C(C=NN2)C=O
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol) 202.213
PubChem CID 737224
Formula Weight 202.21
Percent Purity 95%
Chemical Name or Material 3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.