missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzoyl)propionic acid, 98+%

Product Code. 11493737
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493737 5 g 5g
11403747 25 g 25g
11413747 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493737 Supplier Thermo Scientific Alfa Aesar Supplier No. A18210.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3153-44-4 | C11H12O4 | 208.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-44-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00002795
InChI Key OMTDIBZSUZNVJK-UHFFFAOYSA-N
Synonym 3-4-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxobutanoic acid, 4-4-methoxyphenyl-4-oxobutyric acid, 3-4-methoxybenzoyl propanoic acid, beta-p-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxo-butyric acid, 4-4-methoxyphenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-methoxy-gamma-oxo, maybridge1_002003, acmc-1cl1u
PubChem CID 76613
IUPAC Name 4-(4-methoxyphenyl)-4-oxobutanoic acid
SMILES COC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 785074
Formula Weight 208.21
Percent Purity ≥98%
Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.