missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzoyl)acrylic acid, 98%

Product Code. 11314905
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11314905

Brand: Thermo Scientific Alfa Aesar L18071.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5711-41-1 | C11H10O4 | 206.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5711-41-1
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.20
MDL Number MFCD00020486
InChI Key WORYXBDHTBWLLL-VOTSOKGWSA-N
Synonym 3-4-methoxybenzoyl acrylic acid, 4-4-methoxyphenyl-4-oxobut-2-enoic acid, 2e-4-4-methoxyphenyl-4-oxobut-2-enoic acid, 3-p-methoxybenzoyl acrylic acid, acrylic acid, 3-p-methoxybenzoyl, trans-3-4-methoxybenzoyl acrylic acid, e-4-4-methoxyphenyl-4-oxobut-2-enoic acid, gamma-oxo-4-methoxybenzenecrotonic acid, 4-4-methoxyphenyl-4-oxo-buten-2-saeure, 4-4-methoxyphenyl-4-oxo-2-butenoic acid
PubChem CID 5355095
IUPAC Name (E)-4-(4-methoxyphenyl)-4-oxobut-2-enoic acid
SMILES COC1=CC=C(C=C1)C(=O)\C=C\C(O)=O

Specifications

Melting Point 137°C to 140°C
Quantity 1 g
Beilstein 474529
Formula Weight 206.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxybenzoyl)acrylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.