missing translation for 'onlineSavingsMsg'
Learn More

3-(4’-Methoxy-3’-sulfonamidophenyl)-2-propylamine Hydrochloride, TRC

Product Code. 30483598
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30483598

Brand: Toronto Research Chemicals TRCM26842025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4'-Methoxy-3'-sulfonamidophenyl)-2-propylamine hydrochloride
CAS 86225-65-2
Molecular Formula C10 H16 N2 O3 S . Cl H
Quantity 25 mg
Synonym Benzenesulfonamide','5-(2-aminopropyl)-2-methoxy-','hydrochloride (1:1), Benzenesulfonamide','5-(2-aminopropyl)-2-methoxy-','monohydrochloride (9CI), 3-(4'-Methoxy-3'-sulfonamidophenyl)-2-propylamine hydrochloride, 5-[(2RS)-2-Aminopropyl]-2-methoxybenzenesulfonamide hydrochloride, (±)-5-(2-Aminopropyl)-2-methoxybenzenesulfonamide hydrochloride, 5-(2-Aminopropyl)-2-methoxybenzenesulfonamide hydrochloride
InChI Formula InChI=1S/C10H16N2O3S.ClH/c1-7(11)5-8-3-4-9(15-2)10(6-8)16(12,13)14;/h3-4,6-7H,5,11H2,1-2H3,(H2,12,13,14);1H
SMILES Cl.COc1ccc(CC(C)N)cc1S(=O)(=O)N
IUPAC Name 5-(2-aminopropyl)-2-methoxybenzenesulfonamide;hydrochloride
Molecular Weight (g/mol) 280.77
Formula Weight 280.0648
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.