missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Isopropylidenedioxy) Shikimic Acid Methyl Ester Acetate, TRC

Product Code. 30392921
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392921 100 mg
30397560 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392921 Supplier Toronto Research Chemicals Supplier No. TRCI870400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143308-74-1
Molecular Formula C13H18O6
Molecular Weight (g/mol) 270.28
Synonym (3R,4S,5R)-7-(Acetyloxy)-3a,6,7,7a-tetrahydro-2,2-dimethyl-1,3-benzodioxole-5-carboxylic Acid Methyl Ester, (3R,4S,5R)-5-Hydroxy-3,4-(isopropylidenedioxy)-1-cyclohexene-1-carboxylic Acid Methyl Ester Acetate
IUPAC Name methyl (3aR,7R,7aS)-7-acetyloxy-2,2-dimethyl-3a,6,7,7a-tetrahydro-1,3-benzodioxole-5-carboxylate
SMILES COC(=O)C1=C[C@H]2OC(C)(C)O[C@H]2[C@@H](C1)OC(=O)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18O6/c1-7(14)17-9-5-8(12(15)16-4)6-10-11(9)19-13(2,3)18-10/h6,9-11H,5H2,1-4H3/t9-,10-,11+/m1/s1
Formula Weight 270.1103
Chemical Name or Material 3,4-(Isopropylidenedioxy) Shikimic Acid Methyl Ester Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.