missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionitrile, TRC

Product Code. 30407683
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407683 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407683 Supplier Toronto Research Chemicals Supplier No. TRCH949500500MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17362-17-3
Molecular Formula C9 H9 N O
Quantity 500 mg
Synonym Benzenepropanenitrile, 4-hydroxy-, Hydrocinnamonitrile, p-hydroxy- (6CI,7CI), 4-Hydroxybenzenepropanenitrile, 3-(4-Hydroxyphenyl)propanenitrile, 3-(4-Hydroxyphenyl)propionitrile, 3-(p-Hydroxyphenyl)propionitrile, 4-(2-Cyanoethyl)phenol, 4-Hydroxyhydrocinnamonitrile, NSC 163340, p-(2-Cyanoethyl)phenol, β-(4-Hydroxyphenyl)propionitrile, β-(p-Hydroxyphenyl)propionitrile
InChI Formula InChI=1S/C9H9NO/c10-7-1-2-8-3-5-9(11)6-4-8/h3-6,11H,1-2H2
SMILES Oc1ccc(CCC#N)cc1
IUPAC Name 3-(4-hydroxyphenyl)propanenitrile
Molecular Weight (g/mol) 147.17
Formula Weight 147.0684
Chemical Name or Material 3-(4-Hydroxyphenyl)propionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.