missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionitrile, 98%

Product Code. 11322107
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11322107

Brand: Thermo Scientific Alfa Aesar L07934.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17362-17-3 | C9H9NO | 147.18 g/mol

3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17362-17-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00016492
InChI Key KDMJGLYRWRHKJS-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propionitrile, 3-4-hydroxyphenyl propanenitrile, benzenepropanenitrile, 4-hydroxy, 3-p-hydroxyphenyl propiononitrile, 3-4-hydroxy-phenyl-propionitrile, .beta.-4-hydroxyphenyl propionitrile, pubchem13636, acmc-1bvt5, 4-hydroxyhydrocinnamonitrile, 4-hydroxyphenylpropionitrile
PubChem CID 87079
IUPAC Name 3-(4-hydroxyphenyl)propanenitrile
SMILES OC1=CC=C(CCC#N)C=C1

Specifications

Melting Point 54°C to 58°C
Boiling Point 166°C (8 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 1448680
Solubility Information Insoluble in water.
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 3-(4-Hydroxyphenyl)propionitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.