missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionic Acid, TRC

Product Code. 30134321
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
Unit Size:
100g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134321 100 g 100g
30137726 50 g 50g
30008000 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134321 Supplier Toronto Research Chemicals Supplier No. TRCH950820100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-97-3
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym Benzenepropanoic acid, 4-hydroxy-, Hydrocinnamic acid, p-hydroxy- (7CI,8CI), 4-Hydroxybenzenepropanoic acid, 2,3-Dihydro-p-coumaric acid, 3-(4-Hydroxyphenyl)propanoic acid, 3-(4-Hydroxyphenyl)propionic acid, 3-(4'-Hydroxyphenyl)propionic acid, 3-(p-Hydroxyphenyl)propanoic acid, 3-(p-Hydroxyphenyl)propionic acid, 4-(2-Carboxyethyl)phenol, 4-(2-Carboxylethyl)phenol, 4-Hydroxybenzenepropionic acid, 4-Hydroxydihydrocinnamic acid, Desaminotyrosine, Dihydro-p-coumaric acid, Dihydrocoumaric acid, Hydro-p-coumaric acid, NSC 40949, NSC 65596, Phloretic acid, Phloretinic acid, QuantaBlu, p-Hydroxybenzene propanoic acid, p-Hydroxyhydrocinnamic acid, p-Hydroxyphenylpropionic acid, β-(4-Hydroxyphenyl)propionic acid, β-(p-Hydroxyphenyl)propionic acid
IUPAC Name 3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)CCc1ccc(O)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H10O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5,10H,3,6H2,(H,11,12)
Formula Weight 166.063
Chemical Name or Material Phloretic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.