missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)chroman-7-yl Hexanoate, TRC

Product Code. 30400097
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400097 100 mg 100mg
30389060 25 mg 25mg
30402872 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400097 Supplier Toronto Research Chemicals Supplier No. TRCH918085100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-Hydroxyphenyl)chroman-7-yl Hexanoate
Analyte or Component Names 3-(4-Hydroxyphenyl)chroman-7-yl Hexanoate
Shipping Condition Room Temperature
Molecular Formula C21 H24 O4
InChI Formula InChI=1S/C21H24O4/c1-2-3-4-5-21(23)25-19-11-8-16-12-17(14-24-20(16)13-19)15-6-9-18(22)10-7-15/h6-11,13,17,22H,2-5,12,14H2,1H3
Solution Type Neat
SMILES CCCCCC(=O)Oc1ccc2CC(COc2c1)c3ccc(O)cc3
IUPAC Name [3-(4-hydroxyphenyl)-3,4-dihydro-2H-chromen-7-yl] hexanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 340.413
Quantity 100 mg
Formula Weight 340.167
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.