missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)-1-propanol, TRC

Product Code. 30390530
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390530 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390530 Supplier Toronto Research Chemicals Supplier No. TRCH9528302.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-Hydroxyphenyl)-1-propanol
CAS 10210-17-0
Molecular Formula C9H12O2
Quantity 2.5 g
Synonym 1-(4-Hydroxyphenyl)-3-propanol, 3-(4-Hydroxyphenyl)-1-propanol, 3-(4-Hydroxyphenyl)propanol, 3-(p-Hydroxyphenyl)-1-propanol, 3-(p-Hydroxyphenyl)propyl alcohol, 4-(3-Hydroxypropyl)phenol, 4-Hydroxybenzenepropanol, Dihydro-p-coumaryl Alcohol
InChI Formula InChI=1S/C9H12O2/c10-7-1-2-8-3-5-9(11)6-4-8/h3-6,10-11H,1-2,7H2
SMILES OCCCc1ccc(O)cc1
IUPAC Name 4-(3-hydroxypropyl)phenol
Molecular Weight (g/mol) 152.19
Formula Weight 152.0837
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.