missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)-1,1,3-trimethyl-5-indanol, TRC

Product Code. 30398592
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398592 1 g -
30405754 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398592 Supplier Toronto Research Chemicals Supplier No. TRCH9509351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10527-11-4
Molecular Formula C18 H20 O2
Molecular Weight (g/mol) 268.35
Synonym 1H-Inden-5-ol, 2,3-dihydro-3-(4-hydroxyphenyl)-1,1,3-trimethyl-, 5-Indanol, 3-(p-hydroxyphenyl)-1,1,3-trimethyl- (6CI,7CI,8CI), 2,3-Dihydro-3-(4-hydroxyphenyl)-1,1,3-trimethyl-1H-inden-5-ol, 1,3,3-Trimethyl-1-p-hydroxyphenylindan-6-ol, 3-(4-Hydroxyphenyl)-1,1,3-trimethyl-5-indanol, 3-(p-Hydroxyphenyl)-1,1,3-trimethyl-5-indanol, 5-Hydroxy-3-(4-hydroxyphenyl)-1,1,3-trimethylindan, PIPE-CD
IUPAC Name 3-(4-hydroxyphenyl)-1,1,3-trimethyl-2H-inden-5-ol
SMILES CC1(C)CC(C)(c2ccc(O)cc2)c3cc(O)ccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H20O2/c1-17(2)11-18(3,12-4-6-13(19)7-5-12)16-10-14(20)8-9-15(16)17/h4-10,19-20H,11H2,1-3H3
Formula Weight 268.1463
Chemical Name or Material 3-(4-Hydroxyphenyl)-1,1,3-trimethyl-5-indanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.