missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)cyclopentanone, TRC

Product Code. 30396538
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396538 50 mg -
30408313 5 mg 5mg
30404580 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396538 Supplier Toronto Research Chemicals Supplier No. TRCF40256850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165591-10-6
Molecular Formula C11H11FO
Molecular Weight (g/mol) 178.2
SMILES Fc1ccc(cc1)C2CCC(=O)C2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11FO/c12-10-4-1-8(2-5-10)9-3-6-11(13)7-9/h1-2,4-5,9H,3,6-7H2
Formula Weight 178.0794
Chemical Name or Material 3-(4-Fluorophenyl)cyclopentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.