missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 15488245
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488245 Supplier Thermo Scientific Alfa Aesar Supplier No. H32944.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 306936-57-2
Melting Point 170°C
Molecular Formula C10H7FN2O
MDL Number MFCD01922131
Quantity 1 g
Sensitivity Air Sensitive
Synonym 3-4-fluorophenyl-1h-pyrazole-4-carbaldehyde, 5-4-fluorophenyl-1h-pyrazole-4-carbaldehyde, 3-4-fluorophenyl-1h-pyrazole-4-carboxaldehyde, 3-4-fluorophenyl pyrazole-4-carboxaldehyde, 5-4-fluorophenyl pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-4-fluorophenyl, 3-4-fluorophenyl-2h-pyrazole-4-carbaldehyde, acmc-1ajet, fluorophenylpyrazolecarbaldehyde, pyrazole-4-carboxaldehyde, 3-4-fluorophenyl
InChI Key CMXTUUXWJBLVEH-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=C(C=NN2)C=O)F
IUPAC Name 5-(4-fluorophenyl)-1H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol) 190.177
PubChem CID 605439
Formula Weight 190.18
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.