missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10331735
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10331735 5 g 5g
10723843 1 g 1g
10104104 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10331735 Supplier Thermo Scientific Maybridge Supplier No. SP01479DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-57-2
Molecular Formula C10H7FN2O
Molecular Weight (g/mol) 190.177
MDL Number MFCD01922131
InChI Key CMXTUUXWJBLVEH-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1h-pyrazole-4-carbaldehyde, 5-4-fluorophenyl-1h-pyrazole-4-carbaldehyde, 3-4-fluorophenyl-1h-pyrazole-4-carboxaldehyde, 3-4-fluorophenyl pyrazole-4-carboxaldehyde, 5-4-fluorophenyl pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-4-fluorophenyl, 3-4-fluorophenyl-2h-pyrazole-4-carbaldehyde, acmc-1ajet, fluorophenylpyrazolecarbaldehyde, pyrazole-4-carboxaldehyde, 3-4-fluorophenyl
PubChem CID 605439
IUPAC Name 5-(4-fluorophenyl)-1H-pyrazole-4-carbaldehyde
SMILES C1=CC(=CC=C1C2=C(C=NN2)C=O)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 190.18
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.