missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorobenzoyl)propionic acid, 97%

Product Code. 11474794
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474794 25 g 25g
11464794 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474794 Supplier Thermo Scientific Alfa Aesar Supplier No. A13755.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 366-77-8 | C10H9FO3 | 196.177 g/mol

7-Fluoro-1-tetralone was prepared from 3-(4-fluorobenzoyl)-propionic acid by Wolff-Kishner reduction followed by ring closure with polyphosphoric acid and other substituted 1-tetralones where commercially available. Reduction of 3-(4-fluorobenzoyl)propionic acid with an excess of tert-butylamine borane (TBAB) in the presence of AlCl3 provided 4-(4-fluorophenyl)-1-butanol (87) in 63% yield for the synthesis of LM-1507 sodium salt. Penfluridol 1-[4,4-bis(4-fluorophenyl)butyl]-4-(4-chloro-3-trifluoromethylphenyl)- 4-hydroxypiperidine, C 28 H 27 ClF 5 NO, M r 523.99, mp 105 - 107°C, is obtained by reducing the ketone of 3-(4-fluorobenzoyl)propionic acid into a lactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Fluoro-1-tetralone was prepared from 3-(4-fluorobenzoyl)-propionic acid by Wolff-Kishner reduction followed by ring closure with polyphosphoric acid and other substituted 1-tetralones where commercially available. Reduction of 3-(4-fluorobenzoyl)propionic acid with an excess of tert-butylamine borane (TBAB) in the presence of AlCl3 provided 4-(4-fluorophenyl)-1-butanol (87) in 63% yield for the synthesis of LM-1507 sodium salt. Penfluridol 1-[4,4-bis(4-fluorophenyl)butyl]-4-(4-chloro-3-trifluoromethylphenyl)- 4-hydroxypiperidine, C 28 H 27 ClF 5 NO, M r 523.99, mp 105 - 107°C, is obtained by reducing the ketone of 3-(4-fluorobenzoyl)propionic acid into a lactone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-77-8
Molecular Formula C10H9FO3
Molecular Weight (g/mol) 196.177
MDL Number MFCD00002793
InChI Key WUYWHIAAQYQKPP-UHFFFAOYSA-N
Synonym 3-4-fluorobenzoyl propionic acid, 4-4-fluorophenyl-4-oxobutanoic acid, 3-4-fluorobenzoyl propanoic acid, haloperidol metabolite iii, unii-v2z6n6jc9z, beta-4-fluorobenzoyl propionic acid, v2z6n6jc9z, 3-p-fluorobenzoyl propionic acid, 4-4-fluoro-phenyl-4-oxo-butyric acid
PubChem CID 101359
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
SMILES C1=CC(=CC=C1C(=O)CCC(=O)O)F

Specifications

Melting Point 99°C to 103°C
Odor Odorless
Quantity 25 g
Beilstein 958145
Solubility Information Insoluble in water.
Formula Weight 196.18
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorobenzoyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.