missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluoro-3,5-dimethylphenyl)propionic acid, 97%, Thermo Scientific™

Catalog Number 15487835
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017779-63-3
C11H13FO2
196.221
MFCD09832377
QTKWKNDSBHCYCF-UHFFFAOYSA-N
3-4-fluoro-3,5-dimethylphenyl propionic acid, 3-4-fluoro-3,5-dimethylphenyl propanoic acid, 3-4-fluoro-3,5-dimethylphenyl propionic acid, jrd
46737690
3-(4-fluoro-3,5-dimethylphenyl)propanoic acid
CC1=CC(=CC(=C1F)C)CCC(=O)O
This item is not returnable. View return policy
1017779-63-3
C11H13FO2
196.221
MFCD09832377
QTKWKNDSBHCYCF-UHFFFAOYSA-N
3-4-fluoro-3,5-dimethylphenyl propionic acid, 3-4-fluoro-3,5-dimethylphenyl propanoic acid, 3-4-fluoro-3,5-dimethylphenyl propionic acid, jrd
46737690
3-(4-fluoro-3,5-dimethylphenyl)propanoic acid
CC1=CC(=CC(=C1F)C)CCC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 77.5°C to 79°C
Quantity 250 mg
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 3-(4-Fluoro-3,5-dimethylphenyl)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'