missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene, 99%

Product Code. 15254418 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254418 5 g 5g
15264418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254418 Supplier Thermo Scientific Acros Supplier No. 454930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126213-50-1 | C6H6O2S | 142.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126213-50-1
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.18
InChI Key GKWLILHTTGWKLQ-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
PubChem CID 4421864
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=CSC=C2O1

Specifications

Melting Point 10.5°C to 12.0°C
Density 1.3310g/mL
Boiling Point 193.0°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 5 g
Specific Gravity 1.331
Formula Weight 142.18
Percent Purity 99%
Chemical Name or Material 3,4-Ethylenedioxythiophene
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Ethylenedioxythiophene, 99%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.