missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene, 97%

Product Code. 15483018
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483018 25 g 25g
15473018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483018 Supplier Thermo Scientific Alfa Aesar Supplier No. H56533.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126213-50-1 | C6H6O2S | 142.172 g/mol

3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.

Solubility
Immsicible with water. Miscible with alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126213-50-1
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD02093622
InChI Key GKWLILHTTGWKLQ-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
PubChem CID 4421864
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=CSC=C2O1

Specifications

Density 1.331
Boiling Point 193°C
Flash Point 104°C (219°F)
Refractive Index 1.5765
Quantity 25 g
UN Number UN2810
Solubility Information Immsicible with water. Miscible with alcohol and ether.
Formula Weight 142.17
Percent Purity 97%
Chemical Name or Material 3,4-Ethylenedioxythiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.