missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxybenzoic Acid, TRC

Product Code. 30385637
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385637 Supplier Toronto Research Chemicals Supplier No. TRCE91779525G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Ethylenedioxybenzoic Acid
CAS 4442-54-0
Molecular Formula C9H8O4
Quantity 25 g
Synonym 1,4-Benzodioxan-6-carboxylic Acid (6CI,7CI,8CI), 1,4-Benzodioxane-6-carboxylic Acid, 2,3-Dihydrobenzo[1,4]dioxin-6-carboxylic Acid, 2,3-Dihydrobenzo[1,4]dioxine-6-carboxylic Acid, 2,3-Dihydrobenzo[b][1,4]-dioxin-6-carboxylic Acid, 2,3-Dihydrobenzo[b][1,4]dioxine-6-carboxylic Acid
InChI Formula InChI=1S/C9H8O4/c10-9(11)6-1-2-7-8(5-6)13-4-3-12-7/h1-2,5H,3-4H2,(H,10,11)
SMILES OC(=O)c1ccc2OCCOc2c1
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
Molecular Weight (g/mol) 180.16
Formula Weight 180.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.