missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxycyclohexylmethyl 3,4-Epoxycyclohexaneca, TRC

Product Code. 30384051
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384051 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384051 Supplier Toronto Research Chemicals Supplier No. TRCE58592010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Epoxycyclohexylmethyl 3,4-Epoxycyclohexaneca
CAS 2386-87-0
Molecular Formula C14 H20 O4
Quantity 10 g
Synonym (3,4-Epoxycyclohexane)methyl 3,4-epoxycyclohexylcarboxylate, (3,4-Epoxycyclohexane)methyl 3',4'-epoxycyclohexyl-carboxylate, (3,4-Epoxycyclohexyl)methyl 3,4-epoxycyclohexylcarboxylate, (3',4'-Epoxycyclohexane)methyl 3,4-epoxycyclohexanecarboxylate, (3',4'-Epoxycyclohexylmethyl) 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexanecarboxylic acid (3,4-epoxycyclohexylmethyl) ester, 3,4-Epoxycyclohexanecarboxylic acid (3',4'-epoxycyclohexyl)methyl ester, 3,4-Epoxycyclohexanemethyl 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexenylmethyl 3',4'-epoxycyclohexenecarboxylate, 3,4-Epoxycyclohexylmethyl 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexylmethyl 3',4'-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexylmethyl 3',4'-epoxycyclohexylcarboxylate, 3,4-Epoxycyclohexylmethylene-(3,4-epoxycyclohexane) carboxylate, 7-Oxabicyclo[4.1.0]hept-3-yl-methyl 7-oxabicyclo[4.1.0]heptane-3-carboxylate, 7-Oxabicyclo[4.1.0]heptan-4-ylmethyl 7-oxabicyclo[4.1.0]heptane-4-carboxylate, Chissonox 221 monomer, EPOX, UP 632 monomer, UVA 1500 monomer, 7-Oxabicyclo[4.1.0]heptane-3-carboxylic acid, 7-oxabicyclo[4.1.0]hept-3-ylmethyl ester
InChI Formula InChI=1S/C14H20O4/c15-14(9-2-4-11-13(6-9)18-11)16-7-8-1-3-10-12(5-8)17-10/h8-13H,1-7H2
SMILES O=C(OCC1CCC2OC2C1)C3CCC4OC4C3
IUPAC Name 7-oxabicyclo[4.1.0]heptan-4-ylmethyl 7-oxabicyclo[4.1.0]heptane-4-carboxylate
Molecular Weight (g/mol) 252.31
Formula Weight 252.1362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.