missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxycyclohexanecarboxylate Methyl Ester, TRC

Product Code. 30180105
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180105 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180105 Supplier Toronto Research Chemicals Supplier No. TRCE5859301G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41088-52-2
Molecular Formula C8H12O3
Quantity 1 g
Synonym 1,2-Epoxy-4-methoxycarbonylcyclohexane, 3,4-Cyclohexene oxide-1-carboxylic acid methyl ester, Methyl 3,4-epoxycyclohexane-1-carboxylate, ERL 4140
InChI Formula InChI=1S/C8H12O3/c1-10-8(9)5-2-3-6-7(4-5)11-6/h5-7H,2-4H2,1H3
SMILES COC(=O)C1CCC2OC2C1
IUPAC Name methyl 7-oxabicyclo[4.1.0]heptane-4-carboxylate
Molecular Weight (g/mol) 156.18
Formula Weight 156.0786
Chemical Name or Material 3,4-Epoxycyclohexanecarboxylate Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.