missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylpyrazole, TRC

Product Code. 30379699
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379699 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379699 Supplier Toronto Research Chemicals Supplier No. TRCD478930100MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dimethylpyrazole
CAS 2820-37-3
Molecular Formula C5 H8 N2
Quantity 100 mg
Synonym 1H-Pyrazole, 3,4-dimethyl-, Pyrazole, 3,4(or 4,5)-dimethyl- (7CI), Pyrazole, 3,4-dimethyl- (6CI,8CI), 3,4-Dimethyl-1H-pyrazole, 3,4-Dimethylpyrazole, 4,5-Dimethylpyrazole
InChI Formula InChI=1S/C5H8N2/c1-4-3-6-7-5(4)2/h3H,1-2H3,(H,6,7)
SMILES Cc1c[nH]nc1C
IUPAC Name 3,4-dimethyl-1H-pyrazole
Molecular Weight (g/mol) 96.13
Formula Weight 96.0687
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.