missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylboronic acid, 97%, May contain varying amounts of anhydride, Thermo Scientific™

Product Code. 10290273
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10290273 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10290273 Supplier Thermo Scientific Maybridge Supplier No. AC35928DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55499-43-9
CAS Min % 96.0
Color White to Beige
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Amber Glass Bottle
Physical Form Powder
Assay Percent Range 97%
Molecular Formula C8H11BO2
MDL Number MFCD01009694
Synonym 3,4-dimethylphenyl boronic acid, 3,4-dimethylbenzeneboronic acid, 3,4-dimethylphenyl boranediol, 3,4-dimethylphenylboronicacid, 4-borono-o-xylene, boronic acid, 3,4-dimethylphenyl, pubchem1832, acmc-209lna, 3.4-dimethylbenzeneboronic acid, 3,4-dimethyl phenylboronic acid
InChI Key KDVZJKOYSOFXRV-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1C)B(O)O
IUPAC Name (3,4-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.98
PubChem CID 2734348
Formula Weight 149.98
Percent Purity ≥96.0%
Chemical Name or Material 3, 4-Dimethylphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.