missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylacetic acid, 98%

Product Code. p-7054772
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15459728

Brand: Thermo Scientific Alfa Aesar H63178.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17283-16-8 | C10H11O2 | 163.20 g/mol

3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17283-16-8
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD02664684
InChI Key OTTPBKINJOYJGW-UHFFFAOYSA-M
PubChem CID 296013
IUPAC Name 2-(3,4-dimethylphenyl)acetic acid
SMILES CC1=CC=C(CC([O-])=O)C=C1C

Specifications

Quantity 1 g
Physical Form Solid
Synonym 2-3,4-dimethylphenyl acetic acid, 3,4-dimethylphenylacetic acid, 3,4-dimethyl-phenyl-acetic acid, 3,4-dimethylphenylaceticacid, 3,4-dimethyphenylacetic acid, 3,4-dimethylphenyl acetic acid, benzeneacetic acid, 3,4-dimethyl, 3,4-dimethylbenzenacetic acid, pubchem10440, benzeneacetic acid,3,4-dimethyl
Solubility Information Slightly soluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.