missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylphenyl isocyanate, 97%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
3,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under dry inert gas.
3,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under dry inert gas.
Specifications
Specifications
| Density | 1.051 |
| Boiling Point | 85°C (2 mmHg) |
| Quantity | 25 g |
| Refractive Index | 1.538 |
| UN Number | UN2206 |
| Beilstein | 775149 |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 147.18 |
| Percent Purity | 97% |
| Chemical Name or Material | 3,4-Dimethylphenyl isocyanate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction