missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenol, 99%

Product Code. 10530741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10530741 500 g Glass bottle 500g
10386740 100 g Glass bottle 100g
10793455 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10530741 Supplier Thermo Scientific Acros Supplier No. 116585000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-65-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-65-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002304
InChI Key YCOXTKKNXUZSKD-UHFFFAOYSA-N
Synonym 3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723
PubChem CID 7249
ChEBI CHEBI:39839
IUPAC Name 3,4-dimethylphenol
SMILES CC1=CC=C(O)C=C1C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 63°C to 67°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C6H3OH
Quantity 500 g
Beilstein 06, 480
Merck Index 15, 10279
Formula Weight 122.17
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethylphenol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.