missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Dimethylphenol, 98+%

Product Code. 11483274
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483274 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483274 Supplier Alfa Aesar™ Supplier No. A10599.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
3,4-Dimethylphenol, is used as an anti-Infective agents. It has wide applications in the analytical techniques such as Gas Chromatography, Reversed Phase Chromatography, HPLC and UHPLC.

Solubility
Slightly soluble in water. Less than 1 mg/mL at 68° F.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive.Hygroscopic. Incompatible with bases, acid chlorides, acid anhydrides, and oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 95-65-8
Melting Point 65°C to 68°C
Boiling Point 227°C
Molecular Formula C8H10O
MDL Number MFCD00002304
Quantity 100 g
UN Number UN2261
Beilstein 1099267
Merck Index 14,10082
Synonym 3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723
InChI Key YCOXTKKNXUZSKD-UHFFFAOYSA-N
SMILES CC1=CC=C(O)C=C1C
IUPAC Name 3,4-dimethylphenol
Molecular Weight (g/mol) 122.17
PubChem CID 7249
ChEBI CHEBI:39839
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethylphenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.