missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylhexane, 97%

Product Code. 15440388
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15440388

Brand: Thermo Scientific Alfa Aesar H53498.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 583-48-2 | C8H18 | 114.232 g/mol

3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-48-2
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00039927
InChI Key RNTWWGNZUXGTAX-UHFFFAOYSA-N
Synonym hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso
PubChem CID 11412
IUPAC Name 3,4-dimethylhexane
SMILES CCC(C)C(C)CC

Specifications

Density 0.72
Boiling Point 119°C
Flash Point 6°C (43°F)
Refractive Index 1.403
Quantity 25 g
UN Number UN1262
Beilstein 1718744
Formula Weight 114.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylhexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.