missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzyl chloride, 97%, Thermo Scientific™

Product Code. 10189769
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 102-46-5
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 98°C (12.0 Torr)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H11Cl
Quantity 25 g
Synonym 3,4-dimethylbenzyl chloride, 4-chloromethyl-1,2-dimethylbenzene, benzene, 4-chloromethyl-1,2-dimethyl, benzene, 1-chloromethyl-3,4-dimethyl, 4-chloromethyl-o-xylene, 4-chloromethyl-1,2-dimethyl-benzene, 1-chloromethyl-3,4-dimethylbenzene, acmc-20anej, pubchem14890
InChI Key UBQRAAXAHIKWRI-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)CCl)C
IUPAC Name 4-(chloromethyl)-1,2-dimethylbenzene
Molecular Weight (g/mol) 154.64
PubChem CID 66018
Formula Weight 154.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethylbenzyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethylbenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'