missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzoic acid, 98%

Product Code. 11489526
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489526 500 g 500g
11469526 25 g 25g
11479526 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489526 Supplier Thermo Scientific Alfa Aesar Supplier No. A19260.36
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 619-04-5 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-04-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002524
InChI Key OPVAJFQBSDUNQA-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
PubChem CID 12073
ChEBI CHEBI:64818
IUPAC Name 3,4-dimethylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)C

Specifications

Melting Point 164°C to 167°C
Quantity 500 g
Odor Odorless
Beilstein 907267
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.