missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzenesulfonyl chloride, 98%

Product Code. 11456556
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456556 25 g 25g
11446556 5 g 5g
11466556 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456556 Supplier Thermo Scientific Alfa Aesar Supplier No. B24806.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-30-8 | C8H9ClO2S | 204.668 g/mol

3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-30-8
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00625742
InChI Key DNMQPRPJIWTNAX-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzene-1-sulfonyl chloride, 3,4-dimethylbenzenesulfonylchloride, 3,4-dimethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethyl, 3,4-dimethylphenyl chlorosulfone, ksc562m4t, 3,4-dimethylphenylsulfonyl chloride, 3,4-dimethylbenzenesulphonyl chloride, 3,4-dimethyl benzenesulfonyl chloride
PubChem CID 3689743
IUPAC Name 3,4-dimethylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)C

Specifications

Quantity 25 g
Odor Acrid
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.