missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzaldehyde, 97%

Product Code. 11498336
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498336 5 g 5g
11408346 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498336 Supplier Thermo Scientific Alfa Aesar Supplier No. A15388.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5973-71-7 | C9H10O | 134.178 g/mol

3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00016612
InChI Key POQJHLBMLVTHAU-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
PubChem CID 22278
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)C

Specifications

Density 1.017
Boiling Point 226°C
Odor Almond-like
Refractive Index 1.551
Quantity 5 g
Beilstein 1634270
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol,ether,toluene. Insoluble in water.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylbenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.