missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylaniline, 99+%

Product Code. 10347330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347330 500 g Glass bottle 500g
10427750 5 g Glass bottle 5g
10563321 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10347330 Supplier Thermo Scientific Acros Supplier No. 155285000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 95-64-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-64-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007810
InChI Key DOLQYFPDPKPQSS-UHFFFAOYSA-N
Synonym 3,4-xylidine, 4-amino-o-xylene, 3,4-xylylamine, 3,4-dimethylphenylamine, benzenamine, 3,4-dimethyl, 3,4-dimethylbenzenamine, 3,4-dimethylaminobenzene, 1-amino-3,4-dimethylbenzene, 4-amino-1,2-dimethylbenzene, aniline, 3,4-dimethyl
PubChem CID 7248
ChEBI CHEBI:39901
IUPAC Name 3,4-dimethylaniline
SMILES CC1=CC=C(N)C=C1C

Specifications

Melting Point 49°C to 51°C
CAS Min % 99.4
Color Beige to Brown
CAS Max % 100.0
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Mass or Crystals
Assay Percent Range 99.4% min. (GC)
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1103
Merck Index 15, 10282
Boiling Point 226°C
Solubility Information Solubility in water: <1g/L (24°C). Other solubilities: slightly soluble in chloroform,soluble in alcohol (>100g/L),diethyl ether,,petroleum ether,very soluble in ligroin
Formula Weight 121.18
Percent Purity 99.50%
Chemical Name or Material 3, 4-Dimethylaniline, 99.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.